Please login to your online account to display your discounted pricing

5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine
Assay Percent Range 97%
Molecular Formula C6H2BrF4N
Formula Weight 243.98
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC452570010

  SDS
thermo fisher scientific
AC452570010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC452570050

  SDS
thermo fisher scientific
AC452570050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine
CAS 1262412-30-5
Assay Percent Range 97%
Quantity 1g
Molecular Formula C6H2BrF4N
Formula Weight 243.98