5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine, 97%, ACROS Organics™
Manufacturer:  Thermo Fisher Scientific AC452570050
 View more versions of this product
Catalog No. AC452570050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Specifications
5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine
5g
96% min. (GC)
5-bromo-3-fluoro-2-trifluoromethyl pyridine
C1=C(C(=NC=C1Br)C(F)(F)F)F
243.98
243.98
68.0°C to 70.0°C (25.0 Torr)
1262412-30-5
C6H2BrF4N
ZYPJKTSNQGVERF-UHFFFAOYSA-N
5-bromo-3-fluoro-2-(trifluoromethyl)pyridine
54772300
97%
Chemical Identifiers
1262412-30-5
243.98
5-bromo-3-fluoro-2-trifluoromethyl pyridine
5-bromo-3-fluoro-2-(trifluoromethyl)pyridine
C6H2BrF4N
ZYPJKTSNQGVERF-UHFFFAOYSA-N
54772300
C1=C(C(=NC=C1Br)C(F)(F)F)F
Safety and Handling
Safety and Handling
GHS Signal Word: Warning
SDS Documents