Learn More
5-Bromo-3-fluoro-2-methoxypyridine, 97%, Thermo Scientific™
$67.26 - $67.26
Chemical Identifiers
CAS | 124432-70-8 |
---|---|
Molecular Formula | C6H5BrFNO |
Molecular Weight (g/mol) | 206.014 |
MDL Number | MFCD04112553 |
InChI Key | DEBFRAAJISQBIE-UHFFFAOYSA-N |
PubChem CID | 14837011 |
IUPAC Name | 5-bromo-3-fluoro-2-methoxypyridine |
SMILES | COC1=NC=C(C=C1F)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH64752MD
|
Thermo Scientific Chemicals
H64752MD |
250 mg |
Each for $67.26
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
124432-70-8 | |
206.014 | |
DEBFRAAJISQBIE-UHFFFAOYSA-N | |
5-bromo-3-fluoro-2-methoxypyridine |
C6H5BrFNO | |
MFCD04112553 | |
14837011 | |
COC1=NC=C(C=C1F)Br |
Specifications
124432-70-8 | |
MFCD04112553 | |
13638736 | |
DEBFRAAJISQBIE-UHFFFAOYSA-N | |
5-bromo-3-fluoro-2-methoxypyridine | |
14837011 | |
97% | |
5-Bromo-3-fluoro-2-methoxypyridine |
C6H5BrFNO | |
250 mg | |
2-methoxy-3-fluoro-5-bromopyridine, 5-bromo-3-fluoro-2-methoxy-pyridine, 5-bromo-3-fluoro-2-methoxypyrdine, pubchem10468, acmc-1c0iq, abbypharma ap-10-5273, pyridine,5-bromo-3-fluoro-2-methoxy, pyridine, 5-bromo-3-fluoro-2-methoxy | |
COC1=NC=C(C=C1F)Br | |
206.014 | |
206.01 | |
Solid |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures