missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-fluoro-2-methoxypyridine, 97%

Manufacturer:  Alfa Aesar™ H6475206

 View more versions of this product

Catalog No. AAH6475206



Specifications

Specifications

5-Bromo-3-fluoro-2-methoxypyridine
5g
C6H5BrFNO
13638736
DEBFRAAJISQBIE-UHFFFAOYSA-N
5-bromo-3-fluoro-2-methoxypyridine
14837011
Solid
White
124432-70-8
MFCD04112553
2-methoxy-3-fluoro-5-bromopyridine, 5-bromo-3-fluoro-2-methoxy-pyridine, 5-bromo-3-fluoro-2-methoxypyrdine, pubchem10468, acmc-1c0iq, abbypharma ap-10-5273, pyridine,5-bromo-3-fluoro-2-methoxy, pyridine, 5-bromo-3-fluoro-2-methoxy
COC1=NC=C(C=C1F)Br
206.014
206.01
97%

Chemical Identifiers

124432-70-8
206.014
DEBFRAAJISQBIE-UHFFFAOYSA-N
14837011
COC1=NC=C(C=C1F)Br
C6H5BrFNO
MFCD04112553
2-methoxy-3-fluoro-5-bromopyridine, 5-bromo-3-fluoro-2-methoxy-pyridine, 5-bromo-3-fluoro-2-methoxypyrdine, pubchem10468, acmc-1c0iq, abbypharma ap-10-5273, pyridine,5-bromo-3-fluoro-2-methoxy, pyridine, 5-bromo-3-fluoro-2-methoxy
5-bromo-3-fluoro-2-methoxypyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents