Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-cyano-2-hydroxy-4,6-dimethylpyridine, 98%

$127.18 - $320.95

Chemical Identifiers

Chemical Name or Material 5-Bromo-3-cyano-2-hydroxy-4,6-dimethylpyridine
CAS 23819-87-6
Molecular Formula C8H7BrN2O
Formula Weight 227.06
MDL Number MFCD00052651
Synonym 5-Bromo-2-hydroxy-4,6-dimethylnicotinonitrile; 5-Bromo-2-hydroxy-4,6-dimethyl-3-pyridinecarbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2705803

  SDS
alfa aesar™
H2705803
1g Each for $320.95

AAH27058MD

  SDS
alfa aesar™
H27058MD
250mg Each for $127.18
Description & Specifications

Specifications

Formula Weight 227.06
Chemical Name or Material 5-Bromo-3-cyano-2-hydroxy-4,6-dimethylpyridine
Assay 98%
Molecular Formula C8H7BrN2O
MDL Number MFCD00052651
CAS 23819-87-6
Melting Point ≈256°C
Synonym 5-Bromo-2-hydroxy-4,6-dimethylnicotinonitrile; 5-Bromo-2-hydroxy-4,6-dimethyl-3-pyridinecarbonitrile
Quantity 1g
Odor Odorless
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.