missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals 5-Bromo-3-chloro-2-methoxypyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 449890050
Specifications
5-Bromo-3-chloro-2-methoxypyridine | |
61°C to 63°C | |
C6H5BrClNO | |
5g | |
XCHXNAQRMMXBGN-UHFFFAOYSA-N | |
5-bromo-3-chloro-2-methoxypyridine | |
40427004 | |
97% |
848366-28-9 | |
96% min. (GC) | |
Glass bottle | |
2-methoxy-3-chloro-5-bromopyridine, 5-bromo-3-chloro-2-methoxy-pyridine, pyridine, 5-bromo-3-chloro-2-methoxy, 2-methoxyl-3-chloro-5bromopyridine, 3-chloro-2-methoxy-5-bromopyridine, pubchem1164, acmc-209pww, abbypharma ap-11-5379, 2-methoxy-3-chloro-5-bromo-yridine, pyridine,5-bromo-3-chloro-2-methoxy | |
COC1=NC=C(C=C1Cl)Br | |
222.47 | |
222.47 |
Chemical Identifiers
848366-28-9 | |
222.47 | |
2-methoxy-3-chloro-5-bromopyridine, 5-bromo-3-chloro-2-methoxy-pyridine, pyridine, 5-bromo-3-chloro-2-methoxy, 2-methoxyl-3-chloro-5bromopyridine, 3-chloro-2-methoxy-5-bromopyridine, pubchem1164, acmc-209pww, abbypharma ap-11-5379, 2-methoxy-3-chloro-5-bromo-yridine, pyridine,5-bromo-3-chloro-2-methoxy | |
5-bromo-3-chloro-2-methoxypyridine |
C6H5BrClNO | |
XCHXNAQRMMXBGN-UHFFFAOYSA-N | |
40427004 | |
COC1=NC=C(C=C1Cl)Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO â Research Use Only