missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-3-chloro-2-methoxypyridine, 96%

Manufacturer:  Alfa Aesar™ H5452203

 View more versions of this product

Catalog No. AAH5452203



Description

Description

Applications
5-Bromo-3-chloro-2-methoxypyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Specifications

Specifications

5-Bromo-3-chloro-2-methoxypyridine
1g
C6H5BrClNO
2-methoxy-3-chloro-5-bromo-yridine
COC1=NC=C(C=C1Cl)Br
222.466
222.47
56°C to 58°C
848366-28-9
MFCD04972382
XCHXNAQRMMXBGN-UHFFFAOYSA-N
5-bromo-3-chloro-2-methoxypyridine
40427004
96%

Chemical Identifiers

848366-28-9
222.466
XCHXNAQRMMXBGN-UHFFFAOYSA-N
40427004
COC1=NC=C(C=C1Cl)Br
C6H5BrClNO
MFCD04972382
2-methoxy-3-chloro-5-bromo-yridine
5-bromo-3-chloro-2-methoxypyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C

Documents