missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-(trifluoromethyl)pyridine, TCI America™

$155.84 - $520.98

Chemical Identifiers

CAS 436799-32-5
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD06657686
InChI Key RPFAUCIXZGMCFN-UHFFFAOYSA-N
Synonym 5-bromo-2-trifluoromethyl pyridine, 2-trifluoromethyl-5-bromopyridine, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromopyridine, trifluoromethyl-5-bromo-2-pyridine, 5-bromo-2-trifluoromethyl-pyridine, 5-bromo-2-trifluoromethyl pyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, pubchem2164
PubChem CID 2761197
IUPAC Name 5-bromo-2-(trifluoromethyl)pyridine
SMILES C1=CC(=NC=C1Br)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B34511G View Documents TCI America
B34511G
1g Each for $155.84
 
B34515G View Documents TCI America
B34515G
5g Each for $520.98
 
Specifications

Chemical Identifiers

436799-32-5
225.996
RPFAUCIXZGMCFN-UHFFFAOYSA-N
2761197
C1=CC(=NC=C1Br)C(F)(F)F
C6H3BrF3N
MFCD06657686
5-bromo-2-trifluoromethyl pyridine, 2-trifluoromethyl-5-bromopyridine, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromopyridine, trifluoromethyl-5-bromo-2-pyridine, 5-bromo-2-trifluoromethyl-pyridine, 5-bromo-2-trifluoromethyl pyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, pubchem2164
5-bromo-2-(trifluoromethyl)pyridine

Specifications

78°C
40°C
436799-32-5
MFCD06657686
5-bromo-2-trifluoromethyl pyridine, 2-trifluoromethyl-5-bromopyridine, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromopyridine, trifluoromethyl-5-bromo-2-pyridine, 5-bromo-2-trifluoromethyl-pyridine, 5-bromo-2-trifluoromethyl pyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, pubchem2164
C1=CC(=NC=C1Br)C(F)(F)F
225.996
226.00
≥98.0% (GC)
White-Yellow
1g
C6H3BrF3N
2811
RPFAUCIXZGMCFN-UHFFFAOYSA-N
5-bromo-2-(trifluoromethyl)pyridine
2761197
Crystal-Powder at 20°C
5-Bromo-2-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

TSCA : No