missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442145000

 View more versions of this product

Catalog No. AC442145000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Bromo-2-(trifluoromethyl)pyridine
42.0°C to 43.0°C
436799-32-5
C6H3BrF3N
RPFAUCIXZGMCFN-UHFFFAOYSA-N
5-bromo-2-(trifluoromethyl)pyridine
2761197
97%
78°C
500mg
96% min. (GC)
2-trifluoromethyl-5-bromopyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, 5-bromo-2-trifluoromethyl pyridine, 5-bromo-2-trifluoromethyl-pyridine, pubchem2164, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromo-2-pyridine, trifluoromethyl-5-bromopyridine
C1=CC(=NC=C1Br)C(F)(F)F
226
226

Chemical Identifiers

436799-32-5
226
2-trifluoromethyl-5-bromopyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, 5-bromo-2-trifluoromethyl pyridine, 5-bromo-2-trifluoromethyl-pyridine, pubchem2164, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromo-2-pyridine, trifluoromethyl-5-bromopyridine
5-bromo-2-(trifluoromethyl)pyridine
C6H3BrF3N
RPFAUCIXZGMCFN-UHFFFAOYSA-N
2761197
C1=CC(=NC=C1Br)C(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

GHS Signal Word: Danger

SDS Documents
Product Certifications