Please login to your online account to display your discounted pricing

5-Bromo-2-(trifluoromethyl)pyridine, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine
Assay Percent Range 97%
Molecular Formula C6H3BrF3N
Formula Weight 226
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC442140050

  SDS
thermo fisher scientific
AC442140050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442145000

  SDS
thermo fisher scientific
AC442145000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226
Quantity 5g
Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine
CAS 436799-32-5
Molecular Formula C6H3BrF3N
Assay Percent Range 97%