missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-thiophenesulfonyl Chloride, TCI America™

$59.58 - $352.92

Chemical Identifiers

CAS 55854-46-1
Molecular Formula C4H2BrClO2S2
Molecular Weight (g/mol) 261.532
MDL Number MFCD00084909
InChI Key WGYBIEOLAFYDEC-UHFFFAOYSA-N
Synonym 5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride
PubChem CID 2733924
IUPAC Name 5-bromothiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1)Br)S(=O)(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28781G View Documents TCI America
B28781G
1g Each for $59.58
 
B28785G View Documents TCI America
B28785G
5g Each for $117.64
 
B287825G View Documents TCI America
B287825G
25g Each for $352.92
 
Specifications

Chemical Identifiers

55854-46-1
261.532
WGYBIEOLAFYDEC-UHFFFAOYSA-N
2733924
C1=C(SC(=C1)Br)S(=O)(=O)Cl
C4H2BrClO2S2
MFCD00084909
5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride
5-bromothiophene-2-sulfonyl chloride

Specifications

110°C
41°C
55854-46-1
MFCD00084909
5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride
C1=C(SC(=C1)Br)S(=O)(=O)Cl
261.532
261.53
≥98.0% (GC,T)
Yellow
1g
C4H2BrClO2S2
3261
WGYBIEOLAFYDEC-UHFFFAOYSA-N
5-bromothiophene-2-sulfonyl chloride
2733924
Crystal-Powder at 20°C
5-Bromo-2-thiophenesulfonyl Chloride
Safety and Handling

Safety and Handling

TSCA : No