missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-thiophenemethanol, 95%

Manufacturer:  Alfa Aesar™ H6149314

 View more versions of this product

Catalog No. AAH6149314



Specifications

Specifications

5-Bromo-2-thiophenemethanol
25g
C5H5BrOS
2-thiophenemethanol, 5-bromo
C1=C(SC(=C1)Br)CO
193.058
193.06
95%
Yellow
79387-71-6
MFCD06202626
DBRSLMCLMMFHEF-UHFFFAOYSA-N
(5-bromothiophen-2-yl)methanol
11435467
Liquid

Chemical Identifiers

79387-71-6
193.058
DBRSLMCLMMFHEF-UHFFFAOYSA-N
11435467
C1=C(SC(=C1)Br)CO
C5H5BrOS
MFCD06202626
2-thiophenemethanol, 5-bromo
(5-bromothiophen-2-yl)methanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents