Please login to your online account to display your discounted pricing

5-Bromo-2-thiophenecarboxaldehyde, 97%, ACROS Organics™


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  AC16665EA

 View more versions of this product

Catalog No. AC16665EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 191.05
Quantity 10g
Color Clear yellow to brown
Density 1.61g/mL
Beilstein 17, I, 148
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 5-Bromo-2-thiophenecarboxaldehyde
Packaging Amber glass bottle
Refractive Index 1.6380 to 1.6410 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C5H3BrOS
CAS 4701-17-1
Synonym 5-Bromo-2-thiophenecarbaldehyde
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 5-Bromo-2-thiophenecarboxaldehyde
CAS 4701-17-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H3BrOS
Formula Weight 191.05
Synonym 5-Bromo-2-thiophenecarbaldehyde
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.