Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-(tert-butoxy)pyrimidine, 95%

$110.61 - $283.27

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-(tert-butoxy)pyrimidine
CAS 121487-13-6
Molecular Formula C8H11BrN2O
Formula Weight 231.09
MDL Number MFCD09037471
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2780503

  SDS
alfa aesar™
H2780503
1g Each for $283.27

AAH27805MD

  SDS
alfa aesar™
H27805MD
250mg Each for $110.61
Description & Specifications

Specifications

Formula Weight 231.09
Molecular Formula C8H11BrN2O
Quantity 1g
Melting Point 60° to 63°C
CAS 121487-13-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Health Hazard 1 Warning
Chemical Name or Material 5-Bromo-2-(tert-butoxy)pyrimidine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD09037471
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.