Please login to your online account to display your discounted pricing

5-Bromo-2-pyrazinamine, Tech., Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  MO00943CB

 View more versions of this product

Catalog No. MO00943CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 174
Quantity 250mg
Assay Tech.
CAS Min % 97.0
Synonym 5-Bromo-2-pyrazinamine (IUPAC)
Packaging Amber glass bottle
Chemical Name or Material 5-Bromo-2-pyrazinamine
CAS 59489-71-3
Molecular Formula C4H4BrN3
MDL Number MFCD00235015
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Bromo-2-pyrazinamine
CAS 59489-71-3
CAS Min % 97.0
Purity Grade Technical Grade
Assay Tech.
Molecular Formula C4H4BrN3
Formula Weight 174
MDL Number MFCD00235015
Synonym 5-Bromo-2-pyrazinamine (IUPAC)
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.

SDS & Safety Documents