Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-phenylpyridine, 95%

$569.55 - $569.55

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-phenylpyridine
CAS 27012-25-5
Molecular Formula C11H8BrN
Formula Weight 234.1
MDL Number MFCD04114181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2781603

  SDS
alfa aesar™
H2781603
1g Each for $569.55
Description & Specifications

Specifications

Formula Weight 234.1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C11H8BrN
Melting Point 73° to 75°C
Quantity 1g
MDL Number MFCD04114181
CAS 27012-25-5
Chemical Name or Material 5-Bromo-2-phenylpyridine

Applications
5-Bromo-2-phenylpyridine is used as a ligand in coordination chemistry and reacts with iridium(III) chloride to get in tris-heteroleptic Iridium(III), which finds application as phosphorescent (triplet) emitters in organic light-emitting diodes (OLEDs) due to its self-emitting nature, fast response, wide viewing angle and low power consumption.

Solubility
Soluble in ethyl ether.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.