missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-nitropyridine, 99%, ACROS Organics™

$42.40 - $42.40

Chemical Identifiers

CAS 39856-50-3
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C5H3BrN2O2
Formula Weight 202.99
MDL Number MFCD00160411
Chemical Name or Material 5-Bromo-2-nitropyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC398640010 SDS Thermo Fisher Scientific
AC398640010
Glass bottle 1g Each for $42.40
AC398640050 SDS Thermo Fisher Scientific
AC398640050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

39856-50-3
100.0
C5H3BrN2O2
MFCD00160411
98.5
99%
202.99
5-Bromo-2-nitropyridine

Specifications

39856-50-3
100.0
1g
C5H3BrN2O2
MFCD00160411
98.5
Glass bottle
99%
202.99
5-Bromo-2-nitropyridine

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning