missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-nitrophenol, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6496614

 View more versions of this product

Catalog No. AAH6496614



Description

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in ethyl acetate and methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Specifications

5-Bromo-2-nitrophenol
40°C to 41°C
C6H4BrNO3
25 g
5-bromo-2-nitrophenol, phenol, 5-bromo-2-nitro, 3-bromo-6-nitrophenol, 4-bromo-2-hydroxynitrobenzene, 2-nitro-5-bromophenol, 5-bromo-2-nitro-phenol, phenol,5-bromo-2-nitro, pubchem16073, acmc-1cct0, 5-bromanyl-2-nitro-phenol
DTWHNSNSUBKGTC-UHFFFAOYSA-N
218.006
218.01
Crystalline Solid
27684-84-0
Yellow
MFCD03095027
2329085
Soluble in ethyl acetate and methanol.
C1=CC(=C(C=C1Br)O)[N+](=O)[O-]
13970496
96%

Chemical Identifiers

27684-84-0
218.006
DTWHNSNSUBKGTC-UHFFFAOYSA-N
13970496
C6H4BrNO3
MFCD03095027
5-bromo-2-nitrophenol, phenol, 5-bromo-2-nitro, 3-bromo-6-nitrophenol, 4-bromo-2-hydroxynitrobenzene, 2-nitro-5-bromophenol, 5-bromo-2-nitro-phenol, phenol,5-bromo-2-nitro, pubchem16073, acmc-1cct0, 5-bromanyl-2-nitro-phenol
C1=CC(=C(C=C1Br)O)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only