missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-nitrobenzaldehyde, TCI America™

$24.44 - $73.33

Chemical Identifiers

CAS 20357-20-4
Molecular Formula C7H4BrNO3
Molecular Weight (g/mol) 230.017
MDL Number MFCD00456508
InChI Key UFRVBZVJVRHSNR-UHFFFAOYSA-N
PubChem CID 97233
IUPAC Name 5-bromo-2-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B41921G View Documents TCI America
B41921G
1g Each for $24.44
 
B41925G View Documents TCI America
B41925G
5g Each for $73.33
 
Specifications

Chemical Identifiers

20357-20-4
230.017
UFRVBZVJVRHSNR-UHFFFAOYSA-N
5-bromo-2-nitrobenzaldehyde
C7H4BrNO3
MFCD00456508
97233
C1=CC(=C(C=C1Br)C=O)[N+](=O)[O-]

Specifications

Undesignated
1g
C7H4BrNO3
UFRVBZVJVRHSNR-UHFFFAOYSA-N
5-bromo-2-nitrobenzaldehyde
97233
Crystal-Powder at 20°C
5-Bromo-2-nitrobenzaldehyde
73°C
20357-20-4
MFCD00456508
C1=CC(=C(C=C1Br)C=O)[N+](=O)[O-]
230.017
230.02
≥98.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : CU4875000

TSCA : No