Please login to your online account to display your discounted pricing

5-Bromo-2-(methylthio)pyrimidine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-(methylthio)pyrimidine
CAS 14001-67-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H5BrN2S
Formula Weight 205.08
MDL Number MFCD00187875
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432380010

  SDS
thermo fisher scientific
AC432380010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC432380050

  SDS
thermo fisher scientific
AC432380050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder of Flakes
Assay ≥96.0%
Formula Weight 205.08
Quantity 1g
Color White
Melting Point 64° to 68°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 5-Bromo-2-(methylthio)pyrimidine
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
CAS 14001-67-3
Molecular Formula C5H5BrN2S
MDL Number MFCD00187875
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.