missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-methylpyridine, 98%

$52.35 - $112.19

Chemical Identifiers

CAS 3430-13-5
Molecular Formula C6H6BrN
Formula Weight 172.03
MDL Number MFCD00661170
Synonym 5-Bromo-2-picoline
Chemical Name or Material 5-Bromo-2-methylpyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2708503 SDS Alfa Aesar™
H2708503
1g Each for $112.19
AAH27085MD SDS Alfa Aesar™
H27085MD
250mg Each for $52.35
Specifications Description & Specifications

Chemical Identifiers

3430-13-5
172.03
5-Bromo-2-picoline
C6H6BrN
MFCD00661170
5-Bromo-2-methylpyridine

Specifications

3430-13-5
172.03
32° to 36°C
1g
98%
C6H6BrN
MFCD00661170
5-Bromo-2-picoline
5-Bromo-2-methylpyridine

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger