missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-2-methylindole, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437560050
Specifications
5-Bromo-2-methylindole | |
Glass bottle | |
104.0°C to 106.0°C | |
1075-34-9 | |
100.0 | |
C9H8BrN | |
BJUZAZKEDCDGRW-UHFFFAOYSA-N | |
5-bromo-2-methyl-1H-indole | |
5003968 | |
Powder |
Authentic | |
Tan to Yellow | |
5g | |
97.5 | |
97.5% min. (HPLC) | |
1h-indole, 5-bromo-2-methyl, 1h-indole,5-bromo-2-methyl, 2-methyl-5-bromo-1h-indole, 2-methyl-5-bromoindole, 5-bromo-2-methyl-indole, 5-bromo-2-methylindol, 5-bromo-2-methylindole, acmc-209xly, pubchem9382 | |
CC1=CC2=C(N1)C=CC(=C2)Br | |
210.07 | |
210.07 | |
98% |
Chemical Identifiers
1075-34-9 | |
210.07 | |
1h-indole, 5-bromo-2-methyl, 1h-indole,5-bromo-2-methyl, 2-methyl-5-bromo-1h-indole, 2-methyl-5-bromoindole, 5-bromo-2-methyl-indole, 5-bromo-2-methylindol, 5-bromo-2-methylindole, acmc-209xly, pubchem9382 | |
5-bromo-2-methyl-1H-indole |
C9H8BrN | |
BJUZAZKEDCDGRW-UHFFFAOYSA-N | |
5003968 | |
CC1=CC2=C(N1)C=CC(=C2)Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
W
GHS Signal Word: Warning