Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-methylbenzotrifluoride, 97+%

$122.05 - $439.97

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-methylbenzotrifluoride
CAS 86845-27-4
Molecular Formula C8H6BrF3
Formula Weight 239.04
MDL Number MFCD01631587
Synonym 4-Bromo-2-(trifluoromethyl)toluene; 4-Methyl-3-(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2343103

  SDS
alfa aesar™
B2343103
1g Each for $122.05

AAB2343106

  SDS
alfa aesar™
B2343106
5g Each for $439.97
Description & Specifications

Specifications

Formula Weight 239.04
Molecular Formula C8H6BrF3
MDL Number MFCD01631587
Assay ≥97%
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Quantity 1g
CAS 86845-27-4
Chemical Name or Material 5-Bromo-2-methylbenzotrifluoride
Health Hazard 1 Warning
Synonym 4-Bromo-2-(trifluoromethyl)toluene; 4-Methyl-3-(trifluoromethyl)bromobenzene
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.