missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-methylbenzothiazole, TCI America™

$146.67 - $436.95

Chemical Identifiers

CAS 63837-11-6
Molecular Formula C8H6BrNS
Molecular Weight (g/mol) 228.107
MDL Number MFCD00603096
InChI Key OLQKNZNXLBILDD-UHFFFAOYSA-N
PubChem CID 3017457
IUPAC Name 5-bromo-2-methyl-1,3-benzothiazole
SMILES CC1=NC2=C(S1)C=CC(=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B17985G View Documents TCI America
B17985G
5g Each for $146.67
 
B179825G View Documents TCI America
B179825G
25g Each for $436.95
 
Specifications

Chemical Identifiers

63837-11-6
228.107
OLQKNZNXLBILDD-UHFFFAOYSA-N
5-bromo-2-methyl-1,3-benzothiazole
C8H6BrNS
MFCD00603096
3017457
CC1=NC2=C(S1)C=CC(=C2)Br

Specifications

170°C
79°C
63837-11-6
MFCD00603096
CC1=NC2=C(S1)C=CC(=C2)Br
228.107
228.11
≥98.0% (GC)
White-Yellow
5g
C8H6BrNS
OLQKNZNXLBILDD-UHFFFAOYSA-N
5-bromo-2-methyl-1,3-benzothiazole
3017457
Crystal-Powder at 20°C
5-Bromo-2-methylbenzothiazole
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator