missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-methylbenzoic Acid, TCI America™

$25.97 - $50.42

Chemical Identifiers

CAS 79669-49-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00267350
InChI Key SEENCYZQHCUTSB-UHFFFAOYSA-N
Synonym 2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
PubChem CID 346004
IUPAC Name 5-bromo-2-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)Br)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30505G View Documents TCI America
B30505G
5g Each for $25.97
 
B305025G View Documents TCI America
B305025G
25g Each for $50.42
 
Specifications

Chemical Identifiers

79669-49-1
215.046
SEENCYZQHCUTSB-UHFFFAOYSA-N
346004
CC1=C(C=C(C=C1)Br)C(=O)O
C8H7BrO2
MFCD00267350
2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
5-bromo-2-methylbenzoic acid

Specifications

White-Yellow
5g
C8H7BrO2
2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
CC1=C(C=C(C=C1)Br)C(=O)O
215.046
215.05
≥98.0% (GC,T)
171°C
79669-49-1
MFCD00267350
SEENCYZQHCUTSB-UHFFFAOYSA-N
5-bromo-2-methylbenzoic acid
346004
Crystal-Powder at 20°C
5-Bromo-2-methylbenzoic Acid
Safety and Handling

Safety and Handling

TSCA : No