missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-methylbenzoic acid, 97%

$37.47 - $154.22

Chemical Identifiers

CAS 79669-49-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00267350
InChI Key SEENCYZQHCUTSB-UHFFFAOYSA-N
Synonym 2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
PubChem CID 346004
IUPAC Name 5-bromo-2-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)Br)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6051603 View Documents Alfa Aesar™
H6051603
1g Each for $37.47
 
AAH6051606 View Documents Alfa Aesar™
H6051606
5g Each for $154.22
 
Specifications

Chemical Identifiers

79669-49-1
215.046
SEENCYZQHCUTSB-UHFFFAOYSA-N
346004
CC1=C(C=C(C=C1)Br)C(=O)O
C8H7BrO2
MFCD00267350
2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
5-bromo-2-methylbenzoic acid

Specifications

167°C to 171°C
79669-49-1
MFCD00267350
SEENCYZQHCUTSB-UHFFFAOYSA-N
5-bromo-2-methylbenzoic acid
346004
97%
1g
C8H7BrO2
2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
CC1=C(C=C(C=C1)Br)C(=O)O
215.046
215.05
5-Bromo-2-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger