Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-methylanisole, 97%

$58.15 - $191.36

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-methylanisole
CAS 67868-73-9
Molecular Formula C8H9BrO
Formula Weight 201.06
MDL Number MFCD06797972
Synonym 4-Bromo-2-methoxytoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3039103

  SDS
alfa aesar™
H3039103
1g Each for $58.15

AAH3039106

  SDS
alfa aesar™
H3039106
5g Each for $191.36
Description & Specifications

Specifications

Formula Weight 201.06
Chemical Name or Material 5-Bromo-2-methylanisole
Assay 97%
Boiling Point 102° to 104°C (13mmHg)
Synonym 4-Bromo-2-methoxytoluene
MDL Number MFCD06797972
CAS 67868-73-9
Molecular Formula C8H9BrO
Quantity 1g