missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-2-methoxypyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 376880250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-Bromo-2-methoxypyridine | |
96.0 | |
1.4530g/mL | |
96°C | |
96% min. (GC) | |
C6H6BrNO | |
MFCD01318952 | |
1.453 | |
XADICJHFELMBGX-UHFFFAOYSA-N | |
5-bromo-2-methoxypyridine | |
2734895 | |
97% |
13472-85-0 | |
100.0 | |
80.0°C (12.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5540 to 1.5560 | |
25mL | |
2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408 | |
COC1=NC=C(C=C1)Br | |
188.02 | |
188.02 |
Chemical Identifiers
13472-85-0 | |
188.02 | |
XADICJHFELMBGX-UHFFFAOYSA-N | |
2734895 | |
COC1=NC=C(C=C1)Br |
C6H6BrNO | |
MFCD01318952 | |
2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408 | |
5-bromo-2-methoxypyridine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only