Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-methoxypyridine-3-carboxaldehyde, 99%

$248.62 - $1,066.78

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-methoxypyridine-3-carboxaldehyde
CAS 103058-87-3
Molecular Formula C7H6BrNO2
Formula Weight 216.04
MDL Number MFCD05663521
Synonym 5-Bromo-2-methoxynicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5006206

  SDS
alfa aesar™
H5006206
5g Each for $1,066.78

AAH5006203

  SDS
alfa aesar™
H5006203
1g Each for $248.62
Description & Specifications

Specifications

Formula Weight 216.04
Molecular Formula C7H6BrNO2
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
CAS 103058-87-3
Assay 99%
MDL Number MFCD05663521
Synonym 5-Bromo-2-methoxynicotinaldehyde
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 5-Bromo-2-methoxypyridine-3-carboxaldehyde

Applications
5-Bromo-2-methoxypyridine-3-carboxaldehyde is used as a synthetic building block. It is also used as an pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.