missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-methoxypyridine-3-carboxaldehyde, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC437390010

 View more versions of this product

Catalog No. AC437390010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Bromo-2-methoxypyridine-3-carboxaldehyde
Glass Bottle
102.0°C to 104.0°C
103058-87-3
100.0
C7H6BrNO2
VRNOWEKCASDTFG-UHFFFAOYSA-N
5-bromo-2-methoxypyridine-3-carbaldehyde
1516403
Powder or Crystalline Powder
Authentic
White to Brown
1g
96.0
96% min. (GC)
3-pyridinecarboxaldehyde, 5-bromo-2-methoxy, 3-pyridinecarboxaldehyde,5-bromo-2-methoxy, 5-bromo-2-methoxy-3-pyridinecarboxaldehyde, 5-bromo-2-methoxy-pyridine-3-carbaldehyde, 5-bromo-2-methoxynicotinaldehyde, 5-bromo-2-methoxypyridin-3-carboxaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde 1g, abbypharma ap-10-5904, pubchem9207
COC1=NC=C(C=C1C=O)Br
216.04
216.04
97%

Chemical Identifiers

103058-87-3
216.04
3-pyridinecarboxaldehyde, 5-bromo-2-methoxy, 3-pyridinecarboxaldehyde,5-bromo-2-methoxy, 5-bromo-2-methoxy-3-pyridinecarboxaldehyde, 5-bromo-2-methoxy-pyridine-3-carbaldehyde, 5-bromo-2-methoxynicotinaldehyde, 5-bromo-2-methoxypyridin-3-carboxaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde 1g, abbypharma ap-10-5904, pubchem9207
5-bromo-2-methoxypyridine-3-carbaldehyde
C7H6BrNO2
VRNOWEKCASDTFG-UHFFFAOYSA-N
1516403
COC1=NC=C(C=C1C=O)Br
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Warning

SDS Documents