Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-methoxynicotinic acid, 97%

$250.12 - $1,113.50

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-methoxynicotinic acid
CAS 54916-66-4
Molecular Formula C7H6BrNO3
Formula Weight 232.04
MDL Number MFCD06656771
Synonym 5-Bromo-2-methoxypyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5006503

  SDS
alfa aesar™
H5006503
1g Each for $250.12

AAH5006506

  SDS
alfa aesar™
H5006506
5g Each for $1,113.50
Description & Specifications

Specifications

Formula Weight 232.04
Health Hazard 1 Warning
Synonym 5-Bromo-2-methoxypyridine-3-carboxylic acid
MDL Number MFCD06656771
Chemical Name or Material 5-Bromo-2-methoxynicotinic acid
CAS 54916-66-4
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H6BrNO3
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.