Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-methoxycinnamic acid, 98%

$78.81 - $1,063.65

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-methoxycinnamic acid
CAS 40803-53-0
Molecular Formula C10H9BrO3
Formula Weight 257.09
MDL Number MFCD00017596
Beilstein 3265594
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1779006

 
alfa aesar™
A1779006
5g Each for $78.81

AAA1779022

 
alfa aesar™
A1779022
100g Each for $1,063.65
Description & Specifications

Specifications

Formula Weight 257.09
Chemical Name or Material 5-Bromo-2-methoxycinnamic acid
Assay 98%
Molecular Formula C10H9BrO3
MDL Number MFCD00017596
CAS 40803-53-0
Melting Point 224° to 226°C
Beilstein 3265594
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.