Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-methoxy-beta-nitrostyrene, 97%

$80.61 - $284.78

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-methoxy-beta-nitrostyrene
CAS 175205-14-8
Molecular Formula C9H8BrNO3
Formula Weight 258.07
MDL Number MFCD00068087
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2482403

  SDS
alfa aesar™
B2482403
1g Each for $80.61

AAB2482406

  SDS
alfa aesar™
B2482406
5g Each for $284.78
Description & Specifications

Specifications

Formula Weight 258.07
MDL Number MFCD00068087
Quantity 1g
Melting Point 120° to 122°C
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
Molecular Formula C9H8BrNO3
Assay 97%
CAS 175205-14-8
Chemical Name or Material 5-Bromo-2-methoxy-beta-nitrostyrene
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.