Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-iodopyrimidine, 98%

$64.19 - $163.16

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-iodopyrimidine
CAS 183438-24-6
Molecular Formula C4H2BrIN2
Formula Weight 284.88
MDL Number MFCD01318111
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1745303

  SDS
alfa aesar™
L1745303
1g Each for $64.19

AAL1745306

  SDS
alfa aesar™
L1745306
5g Each for $163.16
Description & Specifications

Specifications

Formula Weight 284.88
Chemical Name or Material 5-Bromo-2-iodopyrimidine
Assay 98%
Molecular Formula C4H2BrIN2
MDL Number MFCD01318111
CAS 183438-24-6
Health Hazard 1 Warning
Melting Point 100° to 102°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.