missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-iodopyridine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC365940050

 View more versions of this product

Catalog No. AC365940050



Description

Description

Specifications

Specifications

5-Bromo-2-iodopyridine
Glass bottle
112°C to 113°C
98%
97.5
97.5% min. (GC)
MFCD00691506
2-iodo-5-bromo pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromopyridine, 5-bromo-2-iodo pyridine, 5-bromo-2-iodo-pyridine, 5-bromo-2-iodopyridine, 5-cromo-2-iodopyridine, acmc-209fv4, pubchem2409, pyridine, 5-bromo-2-iodo
C1=CC(=NC=C1Br)I
283.89
283.89
98%
Authentic
White to Beige
5g
223463-13-6
100.0
C5H3BrIN
20, II, 154
HSNBRDZXJMPDGH-UHFFFAOYSA-N
5-bromo-2-iodopyridine
593690
Crystalline Powder

Chemical Identifiers

223463-13-6
283.89
HSNBRDZXJMPDGH-UHFFFAOYSA-N
593690
C1=CC(=NC=C1Br)I
C5H3BrIN
MFCD00691506
2-iodo-5-bromo pyridine, 2-iodo-5-bromo-pyridine, 2-iodo-5-bromopyridine, 5-bromo-2-iodo pyridine, 5-bromo-2-iodo-pyridine, 5-bromo-2-iodopyridine, 5-cromo-2-iodopyridine, acmc-209fv4, pubchem2409, pyridine, 5-bromo-2-iodo
5-bromo-2-iodopyridine
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents