missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxypyrimidine, 98%

$88.99 - $328.47

Chemical Identifiers

CAS 38353-06-9
Molecular Formula C4H3BrN2O
Molecular Weight (g/mol) 174.985
MDL Number MFCD00233952
InChI Key VTUDATOSQGYWML-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxypyrimidine, 5-bromopyrimidin-2-ol, 5-bromo-2-pyrimidinol, 5-bromo-2-pyrimidinone, 2 1h-pyrimidinone, 5-bromo, 5-bromopyrimidin-2 1h-one, 5-bromo-2 1h-pyrimidinone, 5-bromopyrimidin-2-one, 5-bromo-pyrimidin-2-ol, 2-pyrimidinol, 5-bromo
PubChem CID 101494
IUPAC Name 5-bromo-1H-pyrimidin-2-one
SMILES C1=C(C=NC(=O)N1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1834703 View Documents Alfa Aesar™
L1834703
1g Each for $88.99
 
AAL1834706 View Documents Alfa Aesar™
L1834706
5g Each for $328.47
 
Specifications

Chemical Identifiers

38353-06-9
174.985
VTUDATOSQGYWML-UHFFFAOYSA-N
101494
C1=C(C=NC(=O)N1)Br
C4H3BrN2O
MFCD00233952
5-bromo-2-hydroxypyrimidine, 5-bromopyrimidin-2-ol, 5-bromo-2-pyrimidinol, 5-bromo-2-pyrimidinone, 2 1h-pyrimidinone, 5-bromo, 5-bromopyrimidin-2 1h-one, 5-bromo-2 1h-pyrimidinone, 5-bromopyrimidin-2-one, 5-bromo-pyrimidin-2-ol, 2-pyrimidinol, 5-bromo
5-bromo-1H-pyrimidin-2-one

Specifications

~230°C
38353-06-9
MFCD00233952
5-bromo-2-hydroxypyrimidine, 5-bromopyrimidin-2-ol, 5-bromo-2-pyrimidinol, 5-bromo-2-pyrimidinone, 2 1h-pyrimidinone, 5-bromo, 5-bromopyrimidin-2 1h-one, 5-bromo-2 1h-pyrimidinone, 5-bromopyrimidin-2-one, 5-bromo-pyrimidin-2-ol, 2-pyrimidinol, 5-bromo
C1=C(C=NC(=O)N1)Br
174.985
174.99
5-Bromo-2-hydroxypyrimidine
1g
C4H3BrN2O
742202
VTUDATOSQGYWML-UHFFFAOYSA-N
5-bromo-1H-pyrimidin-2-one
101494
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA