missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxypyrimidine, 98%

Manufacturer:  Alfa Aesar™ L1834703

 View more versions of this product

Catalog No. AAL1834703



Specifications

Specifications

5-Bromo-2-hydroxypyrimidine
1g
C4H3BrN2O
742202
VTUDATOSQGYWML-UHFFFAOYSA-N
5-bromo-1H-pyrimidin-2-one
101494
98%
~230°C
38353-06-9
MFCD00233952
2 1h-pyrimidinone, 5-bromo
C1=C(C=NC(=O)N1)Br
174.985
174.99

Chemical Identifiers

38353-06-9
174.985
VTUDATOSQGYWML-UHFFFAOYSA-N
101494
C1=C(C=NC(=O)N1)Br
C4H3BrN2O
MFCD00233952
2 1h-pyrimidinone, 5-bromo
5-bromo-1H-pyrimidin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents