Please login to your online account to display your discounted pricing

5-Bromo-2-hydroxybenzyl alcohol, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  331480050

 View more versions of this product

Catalog No. AC331480050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder or Flakes
Formula Weight 203.04
Quantity 5g
Color White to light brown
EINECS Number 219-026-0
Melting Point 106° to 111°C
Beilstein 06, 893
Merck Index 12, 1460
CAS Max % 100.0
Packaging Glass bottle
Name Note 98%
Infrared Spectrum Authentic
Chemical Name or Material 5-Bromo-2-hydroxybenzyl alcohol
Assay Percent Range 98%
Molecular Formula C7H7BrO2
Linear Formula BrC6H3(OH)CHOH
MDL Number MFCD00004618
CAS 2316-64-5
Synonym 5-Bromosalicyl alcohol
CAS Min % 97.5

Specifications

Chemical Name or Material 5-Bromo-2-hydroxybenzyl alcohol
Name Note 98%
CAS 2316-64-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7BrO2
Linear Formula BrC6H3(OH)CHOH
Formula Weight 203.04
MDL Number MFCD00004618
Synonym 5-Bromosalicyl alcohol