Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxybenzyl alcohol, 98+%

$118.84 - $1,648.57

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-hydroxybenzyl alcohol
CAS 2316-64-5
Molecular Formula C7H7BrO2
Formula Weight 203.04
MDL Number MFCD00004618
Beilstein 509726
Synonym 5-Bromosalicyl alcohol; Bromosaligenin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1234406

  SDS
alfa aesar™
A1234406
5g Each for $118.84

AAA1234414

  SDS
alfa aesar™
A1234414
25g Each for $461.78

AAA1234422

  SDS
alfa aesar™
A1234422
100g Each for $1,648.57
Description & Specifications

Specifications

Formula Weight 203.04
Chemical Name or Material 5-Bromo-2-hydroxybenzyl alcohol
Assay ≥98%
Molecular Formula C7H7BrO2
MDL Number MFCD00004618
CAS 2316-64-5
Melting Point 108° to 110°C
Beilstein 509726
Synonym 5-Bromosalicyl alcohol; Bromosaligenin
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.