missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxybenzyl alcohol, 98+%

Manufacturer:  Alfa Aesar™ A1234422

 View more versions of this product

Catalog No. AAA1234422


 

Specifications

Specifications

5-Bromo-2-hydroxybenzyl alcohol
≥98%
MFCD00004618
108°C to 110°C
100g
C1=CC(=C(C=C1Br)CO)O
203.035
203.04
2316-64-5
C7H7BrO2
509726
5-Bromosalicyl alcohol; Bromosaligenin
KNKRHSVKIORZQB-UHFFFAOYSA-N
4-bromo-2-(hydroxymethyl)phenol
75342

Chemical Identifiers

2316-64-5
203.035
KNKRHSVKIORZQB-UHFFFAOYSA-N
75342
C7H7BrO2
MFCD00004618
5-Bromosalicyl alcohol; Bromosaligenin
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents