missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxybenzonitrile, 97%

$37.16 - $124.77

Chemical Identifiers

CAS 40530-18-5
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD00143428
InChI Key PVCONXMDUZOPJH-UHFFFAOYSA-N
Synonym 2-hydroxy-5-bromobenzonitrile
PubChem CID 2757014
IUPAC Name 5-bromo-2-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1Br)C#N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5533303 View Documents Alfa Aesar™
H5533303
1g Each for $37.16
 
AAH5533306 View Documents Alfa Aesar™
H5533306
5g Each for $124.77
 
Specifications

Chemical Identifiers

40530-18-5
198.019
PVCONXMDUZOPJH-UHFFFAOYSA-N
2757014
C1=CC(=C(C=C1Br)C#N)O
C7H4BrNO
MFCD00143428
2-hydroxy-5-bromobenzonitrile
5-bromo-2-hydroxybenzonitrile

Specifications

158°C to 163°C
40530-18-5
MFCD00143428
PVCONXMDUZOPJH-UHFFFAOYSA-N
5-bromo-2-hydroxybenzonitrile
2757014
97%
1g
C7H4BrNO
2-hydroxy-5-bromobenzonitrile
C1=CC(=C(C=C1Br)C#N)O
198.019
198.02
5-Bromo-2-hydroxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 254-958-1

TSCA : No

Recommended Storage : Ambient temperatures