missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxybenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5533306

 View more versions of this product

Catalog No. AAH5533306



Description

Description

Specifications

Specifications

5-Bromo-2-hydroxybenzonitrile
5g
C7H4BrNO
2-hydroxy-5-bromobenzonitrile, 4-bromo-2-cyanophenol, 5-bromo-2-hydroxy-benzonitrile, 5-bromo-2-hydroxybenzenecarbonitrile, 5-bromosalicylnitrile, 5-bromosalicylonitrile, acmc-1apuz, benzonitrile, 5-bromo-2-hydroxy, buttpark 20\02-76, pubchem3782
C1=CC(=C(C=C1Br)C#N)O
198.019
198.02
158°C to 163°C
40530-18-5
MFCD00143428
PVCONXMDUZOPJH-UHFFFAOYSA-N
5-bromo-2-hydroxybenzonitrile
2757014
97%

Chemical Identifiers

40530-18-5
198.019
PVCONXMDUZOPJH-UHFFFAOYSA-N
2757014
C1=CC(=C(C=C1Br)C#N)O
C7H4BrNO
MFCD00143428
2-hydroxy-5-bromobenzonitrile, 4-bromo-2-cyanophenol, 5-bromo-2-hydroxy-benzonitrile, 5-bromo-2-hydroxybenzenecarbonitrile, 5-bromosalicylnitrile, 5-bromosalicylonitrile, acmc-1apuz, benzonitrile, 5-bromo-2-hydroxy, buttpark 20\02-76, pubchem3782
5-bromo-2-hydroxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 254-958-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents