missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Bromo-2'-hydroxyacetophenone, 98%

Manufacturer:  Alfa Aesar™ H6197909

 View more versions of this product

Catalog No. AAH6197909



Description

Description

Applications
5′-Bromo-2′-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV), synthesis of {2′-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate, synthesis of N′-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate, preparation of 6-bromochromen-4-one.

Solubility
Soluble in Chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

5'-Bromo-2'-hydroxyacetophenone
56°C to 60°C
1450-75-5
MFCD00191850
HQCCNFFIOWYINW-UHFFFAOYSA-N
1-(5-bromo-2-hydroxyphenyl)ethanone
95991
Solid
White to Yellow
10g
C8H7BrO2
1-5-bromo-2-hydroxyphenyl ethan-1-one
CC(=O)C1=C(C=CC(=C1)Br)O
215.046
215.04
98%

Chemical Identifiers

1450-75-5
215.046
HQCCNFFIOWYINW-UHFFFAOYSA-N
95991
CC(=O)C1=C(C=CC(=C1)Br)O
C8H7BrO2
MFCD00191850
1-5-bromo-2-hydroxyphenyl ethan-1-one
1-(5-bromo-2-hydroxyphenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : KM5556350

Documents