Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxy-3-nitropyridine, 97%


Quantity 1g

Manufacturer: alfa aesar™  L1985003

 View more versions of this product

Catalog No. AAL1985003

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 219
Chemical Name or Material 5-Bromo-2-hydroxy-3-nitropyridine
Health Hazard 1 Warning
Molecular Formula C5H3BrN2O3
MDL Number MFCD00023473
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 15862-34-7
Assay 97%
Quantity 1g
Beilstein 383853
Odor Nearly Odorless
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 245° to 248°C

Specifications

Chemical Name or Material 5-Bromo-2-hydroxy-3-nitropyridine
CAS 15862-34-7
Molecular Formula C5H3BrN2O3
Formula Weight 219
MDL Number MFCD00023473
Beilstein 383853
Accessories

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents