Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxy-3-nitropyridine, 97%

$38.03 - $106.66

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-hydroxy-3-nitropyridine
CAS 15862-34-7
Molecular Formula C5H3BrN2O3
Formula Weight 219
MDL Number MFCD00023473
Beilstein 383853
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1985003

 
alfa aesar™
L1985003
1g Each for $38.03

AAL1985006

 
alfa aesar™
L1985006
5g Each for $106.66
Description & Specifications

Specifications

Formula Weight 219
Chemical Name or Material 5-Bromo-2-hydroxy-3-nitropyridine
Molecular Formula C5H3BrN2O3
MDL Number MFCD00023473
CAS 15862-34-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Melting Point 245° to 248°C
Beilstein 383853
Quantity 1g
Odor Nearly Odorless
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.