missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-hydroxy-3-methylpyridine, TCI America™

$24.44 - $94.72

Chemical Identifiers

CAS 89488-30-2
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
InChI Key HXPMRPRBABWPKL-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxy-3-methylpyridine, 5-bromo-3-methylpyridin-2-ol, 5-bromo-2-hydroxy-3-picoline, 5-bromo-3-methylpyridin-2 1h-one, 5-bromo-3-methyl-pyridin-2-ol, 5-bromo-3-methyl-2-pyridinol, 2-hydroxy-5-bromo-3-picoline, 2-hydroxy-3-methyl-5-bromopyridine, 5-bromo-3-methyl-2-pyridone, 2 1h-pyridinone, 5-bromo-3-methyl
PubChem CID 285436
IUPAC Name 5-bromo-3-methyl-1H-pyridin-2-one
SMILES CC1=CC(=CNC1=O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B27971G View Documents TCI America
B27971G
1g Each for $24.44
 
B27975G View Documents TCI America
B27975G
5g Each for $94.72
 
Specifications

Chemical Identifiers

89488-30-2
188.024
5-bromo-2-hydroxy-3-methylpyridine, 5-bromo-3-methylpyridin-2-ol, 5-bromo-2-hydroxy-3-picoline, 5-bromo-3-methylpyridin-2 1h-one, 5-bromo-3-methyl-pyridin-2-ol, 5-bromo-3-methyl-2-pyridinol, 2-hydroxy-5-bromo-3-picoline, 2-hydroxy-3-methyl-5-bromopyridine, 5-bromo-3-methyl-2-pyridone, 2 1h-pyridinone, 5-bromo-3-methyl
5-bromo-3-methyl-1H-pyridin-2-one
C6H6BrNO
HXPMRPRBABWPKL-UHFFFAOYSA-N
285436
CC1=CC(=CNC1=O)Br

Specifications

Yellow
1g
C6H6BrNO
HXPMRPRBABWPKL-UHFFFAOYSA-N
5-bromo-3-methyl-1H-pyridin-2-one
285436
Crystal-Powder at 20°C
5-Bromo-2-hydroxy-3-methylpyridine
172°C
89488-30-2
5-bromo-2-hydroxy-3-methylpyridine, 5-bromo-3-methylpyridin-2-ol, 5-bromo-2-hydroxy-3-picoline, 5-bromo-3-methylpyridin-2 1h-one, 5-bromo-3-methyl-pyridin-2-ol, 5-bromo-3-methyl-2-pyridinol, 2-hydroxy-5-bromo-3-picoline, 2-hydroxy-3-methyl-5-bromopyridine, 5-bromo-3-methyl-2-pyridone, 2 1h-pyridinone, 5-bromo-3-methyl
CC1=CC(=CNC1=O)Br
188.024
188.02
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No