missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxy-3-methylpyridine, 98%

Manufacturer:  Alfa Aesar™ H6425306

 View more versions of this product

Catalog No. AAH6425306



Specifications

Specifications

5-Bromo-2-hydroxy-3-methylpyridine
170°C to 174°C (literature)
89488-30-2
MFCD03427657
5-bromo-2-hydroxy-3-methylpyridine, 5-bromo-3-methylpyridin-2-ol, 5-bromo-2-hydroxy-3-picoline, 5-bromo-3-methylpyridin-2 1h-one, 5-bromo-3-methyl-pyridin-2-ol, 5-bromo-3-methyl-2-pyridinol, 2-hydroxy-5-bromo-3-picoline, 2-hydroxy-3-methyl-5-bromopyridine, 5-bromo-3-methyl-2-pyridone, 2 1h-pyridinone, 5-bromo-3-methyl
CC1=CC(=CNC1=O)Br
188.024
188.02
98%
White
5g
C6H6BrNO
1447443
HXPMRPRBABWPKL-UHFFFAOYSA-N
5-bromo-3-methyl-1H-pyridin-2-one
285436
Solid

Chemical Identifiers

89488-30-2
188.024
HXPMRPRBABWPKL-UHFFFAOYSA-N
285436
CC1=CC(=CNC1=O)Br
C6H6BrNO
MFCD03427657
5-bromo-2-hydroxy-3-methylpyridine, 5-bromo-3-methylpyridin-2-ol, 5-bromo-2-hydroxy-3-picoline, 5-bromo-3-methylpyridin-2 1h-one, 5-bromo-3-methyl-pyridin-2-ol, 5-bromo-3-methyl-2-pyridinol, 2-hydroxy-5-bromo-3-picoline, 2-hydroxy-3-methyl-5-bromopyridine, 5-bromo-3-methyl-2-pyridone, 2 1h-pyridinone, 5-bromo-3-methyl
5-bromo-3-methyl-1H-pyridin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents