Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxy-3-methylpyridine, 98%

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-hydroxy-3-methylpyridine
CAS 89488-30-2
Formula Weight 188.02
MDL Number MFCD03427657
Beilstein 1447443
Synonym 5-Bromo-2-hydroxy-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6425306

  SDS
alfa aesar™
H6425306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6425314

  SDS
alfa aesar™
H6425314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6425322

  SDS
alfa aesar™
H6425322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 170° to 174°C (lit.)
Formula Weight 188.02
Chemical Name or Material 5-Bromo-2-hydroxy-3-methylpyridine
Color Off-white
Physical Form Solid
Assay 98%
Quantity 5g
MDL Number MFCD03427657
CAS 89488-30-2
Beilstein 1447443
Synonym 5-Bromo-2-hydroxy-3-picoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.