Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol

$124.68 - $434.08

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol
CAS 35090-64-3
Molecular Formula C8H9BrO3
Formula Weight 233.06
MDL Number MFCD02081986
Synonym 4-Bromo-2-hydroxymethyl-6-methoxyphenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5056903

 
alfa aesar™
H5056903
1g Each for $434.08

AAH50569MD

 
alfa aesar™
H50569MD
250mg Each for $124.68
Description & Specifications

Specifications

Formula Weight 233.06
Chemical Name or Material 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol
Molecular Formula C8H9BrO3
MDL Number MFCD02081986
CAS 35090-64-3
Synonym 4-Bromo-2-hydroxymethyl-6-methoxyphenol
Quantity 1g