Please login to your online account to display your discounted pricing

5-Bromo-2-hydroxy-3-methoxybenzaldehyde, 97%, Maybridge


Quantity 100g

Manufacturer: fisher scientific company  BTB03758FA

 View more versions of this product

Catalog No. BTB03758FA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 231.04
Quantity 100g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 5-Bromo-2-hydroxy-3-methoxybenzaldehyde
Assay Percent Range 97%
Molecular Formula C8H7BrO3
Linear Formula CH3OC6H2(OH)(Br)CH(O)
MDL Number MFCD00016593
CAS 5034-74-2
CAS Min % 96.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Bromo-2-hydroxy-3-methoxybenzaldehyde
CAS 5034-74-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7BrO3
Linear Formula CH3OC6H2(OH)(Br)CH(O)
Formula Weight 231.04
MDL Number MFCD00016593
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.